Learn More
Biolegend PE anti-human Lymphotoxin β Receptor (LT-βR)
Supplier:  Biolegend 322008
Encompass_Preferred
PE anti-human Lymphotoxin β Receptor (LT-βR) [31G4D8]; Isotype: Mouse IgG2b, κ; Reactivity: Human, African Green, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests
Catalog No. 50-167-107