Learn More
Biolegend PE anti-human CD300c
Supplier:  Biolegend 334804
Encompass_Preferred
PE anti-human CD300c [TX45]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests
Catalog No. 50-167-811