Learn More
Biolegend PE anti-Human Podoplanin
Supplier:  Biolegend 337004
Encompass_Preferred
PE anti-Human Podoplanin [NC-08]; Isotype: Rat IgG2a, λ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 50-167-883