Learn More
Biolegend PE anti-human PSMA (FOLH1)
Supplier:  Biolegend 342503
Encompass_Preferred
PE anti-human PSMA (FOLH1) [LNI-17]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 25 tests
Catalog No. 50-604-268