Learn More
Biolegend PE anti-human TCR α/β
Supplier:  Biolegend 306707
Encompass_Preferred
PE anti-human TCR α/β [IP26]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 25 tests
Catalog No. 50-166-379