Learn More
Biolegend PE anti-human TCR Vβ13.2
Supplier:  Biolegend 333108
Encompass_Preferred
PE anti-human TCR Vβ13.2 [H132]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 50-167-711