Learn More
Biolegend PE anti-human TCR Vβ9
Supplier:  Biolegend 349204
Encompass_Preferred
PE anti-human TCR Vβ9 [MKB1]; Isotype: Mouse IgG2b, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 50-168-511