Learn More
Biolegend PE anti-human TCR Vγ9
Supplier:  Biolegend 331307
Encompass_Preferred
PE anti-human TCR Vγ9 [B3]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Chimpanzee, Cynomolgus; Apps: FC; Size: 25 tests
Catalog No. 50-167-637