Learn More
Biolegend PE anti-human TRA-1-60-R
Supplier:  Biolegend 330609
Encompass_Preferred
PE anti-human TRA-1-60-R [TRA-1-60-R]; Isotype: Mouse IgM, κ; Reactivity: Human; Apps: FC; Size: 25 tests
Catalog No. 50-167-586