Learn More
Biolegend PE Anti-Human/Mouse Granzyme B Recombinant Antibody | 25 Tests
Supplier:  Biolegend 372207
Encompass_Preferred
Granzyme B is a 32 kD serine protease, also known as granzyme-2, serine protease B, CCP1, Asp-ase, and CTLA-1. Granzyme B is abundantly stored in the granules of cytotoxic T lymphocytes and NK cells. Low level of expression has been reported in granulocytes, B cells, and activated dendritic cells. Granzyme B is crucial for rapid induction of cell death and apoptosis through interaction with mannose-6-phosphate receptor.
Catalog No. NC1694242