Learn More
Biolegend PE anti-human β2-microglobulin
Supplier:  Biolegend 395704
Encompass_Preferred
PE anti-human β2-microglobulin [A17082A]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. NC1794263