Learn More
Biolegend PE anti-human CD110
Supplier:  Biolegend 393804
Encompass_Preferred
PE anti-human CD110 [S16017A]; Isotype: Mouse IgG2b, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 50-207-9050