Learn More
Biolegend PE anti-human CD171 (L1CAM)
Supplier:  Biolegend 371604
Encompass_Preferred
PE anti-human CD171 (L1CAM) [L1-OV198.5]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 50-207-8671