Learn More
Biolegend PE anti-human CD186 (CXCR6)
Supplier:  Biolegend 356003
Encompass_Preferred
PE anti-human CD186 (CXCR6) [K041E5]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: FC; Size: 25 tests
Catalog No. 50-207-7451