Learn More
Biolegend PE anti-human CD22
Supplier:  Biolegend 363503
Encompass_Preferred
PE anti-human CD22 [S-HCL-1]; Isotype: Mouse IgG2b, κ; Reactivity: Human; Apps: FC; Size: 25 tests
Catalog No. 50-103-0822