Learn More
Biolegend PE anti-human CD244 (2B4)
Supplier:  Biolegend 393507
Encompass_Preferred
PE anti-human CD244 (2B4) [2-69]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: FC; Size: 25 tests
Catalog No. 50-207-9022