Learn More
Biolegend PE anti-human CD27
Supplier:  Biolegend 356406
Encompass_Preferred
PE anti-human CD27 [M-T271]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests
Catalog No. 50-604-455