Learn More
Biolegend PE anti-human CD27
Supplier:  Biolegend 302807
Encompass_Preferred
PE anti-human CD27 [O323]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus, Squirrel Monkey; Apps: FC; Size: 25 tests
Catalog No. NC1732700