Learn More
Biolegend PE anti-human CD27 Recombinant [QA17A18]; Isotype: Mouse IgG1, ; Reactivity: Human; Apps: FC; Size: 100 tests
Supplier:  Biolegend 393212
Encompass_Preferred
PE anti-human CD27 Recombinant [QA17A18]; Isotype: Mouse IgG1, ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 50-207-8980