Learn More
Biolegend PE anti-human CD274 (B7-H1, PD-L1)
Supplier:  Biolegend 374511
Encompass_Preferred
PE anti-human CD274 (B7-H1, PD-L1) [MIH3]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 25 tests
Catalog No. 50-207-8811