Learn More
Biolegend PE anti-human CD282 (TLR2)
Supplier:  Biolegend 392305
Encompass_Preferred
PE anti-human CD282 (TLR2) [W15145C]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: FC; Size: 25 tests
Catalog No. 50-207-8892