Learn More
Biolegend PE anti-human CD309 (VEGFR2)
Supplier:  Biolegend 393004
Encompass_Preferred
PE anti-human CD309 (VEGFR2) [A16085H]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 50-207-8951