Learn More
Biolegend PE anti-human CD326 (EpCAM) Recombinant [QA20A75]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 25 tests
Catalog No. 502723206
Encompass_Preferred
PE anti-human CD326 (EpCAM) Recombinant [QA20A75]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 25 tests
Catalog No. 50-272-3206 Supplier Biolegend Supplier No. 377507
PE anti-human CD326 (EpCAM) Recombinant [QA20A75]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 25 tests