Learn More
Biolegend PE anti-human CD357 (GITR)
Supplier:  Biolegend 371204
Encompass_Preferred
PE anti-human CD357 (GITR) [108-17]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. NC1439491