Learn More
Biolegend PE anti-human CD357 (GITR)
Supplier:  Biolegend 311604
Encompass_Preferred
PE anti-human CD357 (GITR) [621]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests
Catalog No. NC0524634