Learn More
Cytek Biosciences Inc Antibodies PE Anti-Human CD38 (HIT2) ..... Cat. No. 50-0389-T025 ..... Size 25 tests
Supplier:  Cytek Biosciences Inc 500389T025
Encompass_Preferred
Antibodies PE Anti-Human CD38 (HIT2) ..... Cat. No. 50-0389-T025 ..... Size 25 testsThe HIT2 antibody is specific for human CD38, a 45 kD type II transmembrane glycoprotein expressed on thymocytes, activated T and B cells, and monocytes as well as other non-hematopoietic cells. CD38 is an ectoenzyme that functions to catalyze the synthesis and hydrolysis of cyclic ADP-ribose and is involved in cell signaling, regulating cell adhesion, and activation. Additionally, CD38 has been shown to be a prognostic marker for some leukemias and other diseases.
Catalog No. 50-210-3396