Learn More
Biolegend PE anti-human CD45 Recombinant
Supplier:  Biolegend 384408
Encompass_Preferred
PE anti-human CD45 Recombinant [QA21A24]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 50-253-7850