Learn More
Cytek Biosciences Inc Antibodies PE Anti-Human CD54 (ICAM-1) (15.2) ..... Cat. No. 50-0549-T025 ..... Size 25 tests
Supplier:  Cytek Biosciences Inc 500549T025
Encompass_Preferred
Antibodies PE Anti-Human CD54 (ICAM-1) (15.2) ..... Cat. No. 50-0549-T025 ..... Size 25 testsThe 15.2 antibody reacts with human CD54, also known as ICAM-1 (Intercellular Adhesion Molecule 1), a 90-110 kDa cell surface glycoprotein that is inducibly expressed on both immune and endothelial cells. As its name implies, ICAM-1 participates in cell-cell adhesion between leukocytes and endothelial cells, facilitating leukocyte recruitment and transmigration at sites of inflammation. The ligands for ICAM-1 are also expressed on leukocyte and endothelial cells, and include Mac-1, fibrinogen, and a member of the integrin protein family, LFA-1 (CD11a).The 15.2 antibody may be used for analysis of ICAM-1 expression in human cells and tissues, and is reported to be cross-reactive with porcine ICAM-1.
Catalog No. 50-210-3407