Learn More
Biolegend PE anti-human CD64
Supplier:  Biolegend 399503
Encompass_Preferred
PE anti-human CD64 [S18012C]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 25 tests
Catalog No. 50-207-9410