Learn More
Biolegend PE anti-human CD69
Catalog No. 502537847
Encompass_Preferred
PE anti-human CD69 [W19310K]; Isotype: Rat IgG2a, κ; Reactivity: Human; Apps: FC; Size: 25 tests
Catalog No. 50-253-7847 Supplier Biolegend Supplier No. 388603
PE anti-human CD69 [W19310K]; Isotype: Rat IgG2a, κ; Reactivity: Human; Apps: FC; Size: 25 tests