Learn More
Cytek Biosciences Inc PE aHu CD80 Anti Primary
Supplier: Cytek Biosciences Inc 500809T025
Product Name: PE Anti-Human CD80 (B7-1) (2D10.4)
Product Class: Antibody
Type: Primary
Product Name: PE Anti-Human CD80 (B7-1) (2D10.4)
Cytek Catalog Number: 50-0809-T025
Marker: CD80 (B7-1)
Target Species: Human
Clonality: Monoclonal
Clone: 2D10.4
Host Species: Mouse
Isotype: IgG1, k
Conjugation: PE
Size: 25 Tests
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.