Learn More
ABclonal Technology PE anti-Human CD87/PLAUR
SDP
Catalog No. 5000010987
Encompass_Preferred
This gene encodes the receptor for urokinase plasminogen activator and given its role in localizing and promoting plasmin formation likely influences many normal and pathological processes related to cell-surface plasminogen activation and localized degradation of the extracellular matrix It binds both the proprotein and mature forms of urokinase plasminogen activator and permits the activation of the receptor-bound pro-enzyme by plasmin The protein lacks transmembrane or cytoplasmic domains and may be anchored to the plasma membrane by a glycosyl-phosphatidylinositol (GPI) moiety following cleavage of the nascent polypeptide near its carboxy-terminus However a soluble protein is also produced in some cell types Alternative splicing results in multiple transcript variants encoding different isoforms The proprotein experiences several post-translational cleavage reactions that have not yet been fully defined
Catalog No. 50-000-10987 Supplier ABclonal Technology Supplier No. A26640500T
This gene encodes the receptor for urokinase plasminogen activator and given its role in localizing and promoting plasmin formation likely influences many normal and pathological processes related to cell-surface plasminogen activation and localized degradation of the extracellular matrix It binds both the proprotein and mature forms of urokinase plasminogen activator and permits the activation of the receptor-bound pro-enzyme by plasmin The protein lacks transmembrane or cytoplasmic domains and may be anchored to the plasma membrane by a glycosyl-phosphatidylinositol (GPI) moiety following cleavage of the nascent polypeptide near its carboxy-terminus However a soluble protein is also produced in some cell types Alternative splicing results in multiple transcript variants encoding different isoforms The proprotein experiences several post-translational cleavage reactions that have not yet been fully defined