Learn More
Biolegend PE anti-human CXCL16
Supplier:  Biolegend 360803
Encompass_Preferred
PE anti-human CXCL16 [22-19-12]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 25 tests
Catalog No. 50-207-7948