Learn More
Biolegend PE anti-human CXCL9 (MIG)
Catalog No. 502077724
Encompass_Preferred
PE anti-human CXCL9 (MIG) [J1015E10]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: ICFC; Size: 100 tests
Catalog No. 50-207-7724 Supplier Biolegend Supplier No. 357904
Add to Cart
Add to Cart
PE anti-human CXCL9 (MIG) [J1015E10]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: ICFC; Size: 100 tests