Learn More
Biolegend PE anti-human Granzyme A
Catalog No. 502796500
Encompass_Preferred
PE anti-human Granzyme A [CB9]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: ICFC; Size: 500 μL
This item is not returnable. View return policy
Catalog No. 50-279-6500 Supplier Biolegend Supplier No. 989202
PE anti-human Granzyme A [CB9]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: ICFC; Size: 500 μL