Learn More
Cytek Biosciences Inc Antibodies PE Anti-Human HLA-DR (L243) ..... Cat. No. 50-9952-T500 ..... Size 500 tests
Supplier:  Cytek Biosciences Inc 509952T500
Encompass_Preferred
Antibodies PE Anti-Human HLA-DR (L243) ..... Cat. No. 50-9952-T500 ..... Size 500 testsThe L243 antibody reacts with a member of the human MHC Class II antigens, HLA-DR. The HLA-DR antigen is expressed on B lymphocytes, activated T lymphocytes, activated NK cells, monocytes, macrophages, other antigen presenting cells and progenitor cells. The L243 antibody is specific to an epitope on the alpha subunit of the heterodimeric HLA-DR protein and binds a different epitope than the LN3 antibody clone. It does not cross-react with HLA-DP or HLA-DQ. This antibody is reported to be cross-reactive with non-human primates including Chimpanzee, Cynomolgus, Rhesus and Baboon. Please choose the appropriate format for each application.
Catalog No. 50-210-3461