Learn More
ABclonal Technology PE anti-Human ICOS/CD278
SDP
Supplier:  ABclonal Technology A26444100T
Encompass_Preferred
The protein encoded by this gene belongs to the CD28 and CTLA-4 cell-surface receptor family It forms homodimers and plays an important role in cell-cell signaling immune responses and regulation of cell proliferation
Catalog No. 50-000-10774