Learn More
Cytek Biosciences Inc Antibodies PE Anti-Human IL-4 (MP4-25D2) ..... Cat. No. 50-7048-T025 ..... Size 25 tests
Supplier:  Cytek Biosciences Inc 507048T025
Encompass_Preferred
Antibodies PE Anti-Human IL-4 (MP4-25D2) ..... Cat. No. 50-7048-T025 ..... Size 25 testsThe MP4-25D2 antibody is specific for human IL-4, a pleiotropic cytokine that Th2 cell differentiation, B cell proliferation and differentiation, and B cell class switching to IgE. MP4-25D2 is a neutralizing antibody and is reported to cross react with rhesus monkey IL-4.
Catalog No. 50-210-3453