Learn More
Biolegend PE anti-human MERTK
Supplier:  Biolegend 367608
Encompass_Preferred
PE anti-human MERTK [590H11G1E3]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. NC1183483