Learn More
Biolegend PE anti-human/mouse/rat MR1
Supplier:  Biolegend 396105
Encompass_Preferred
PE anti-human/mouse/rat MR1 [8F2.F9]; Isotype: Mouse IgG1, κ; Reactivity: Human, Mouse, Rat, Cow; Apps: FC; Size: 25 tests
Catalog No. 50-207-9219