Learn More
Biolegend PE anti-MEK1 Phospho (Ser298)
Supplier:  Biolegend 610603
Encompass_Preferred
PE anti-MEK1 Phospho (Ser298) [A16117B]; Isotype: Mouse IgG1, κ; Reactivity: Human, Mouse; Apps: ICFC; Size: 25 tests
Catalog No. 50-208-6692