Learn More
Biolegend PE anti-mouse Blimp-1
Supplier:  Biolegend 150005
Encompass_Preferred
PE anti-mouse Blimp-1 [5E7]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: ICFC; Size: 25 ug
Catalog No. 50-207-1908