Learn More
Cytek Biosciences Inc Antibodies PE Anti-Mouse CD115 (c-fms) (AFS98) ..... Cat. No. 50-1152-U025 ..... Size 25 µg
Supplier:  Cytek Biosciences Inc 501152U025
Encompass_Preferred
Antibodies PE Anti-Mouse CD115 (c-fms) (AFS98) ..... Cat. No. 50-1152-U025 ..... Size 25 µgThe AFS98 antibody is specific for mouse CD115, also known as Colony-Stimulating Factor-1 Receptor (CSF-1R), a 145 kDa receptor from the PDGF receptor family. Receptor activation by the ligands IL-34 or CSF-1 (M-CSF) occurs via homodimerization of CD115 and subsequent tyrosine phosphorylation and ubiquitination of intracellular domains. CD115 signaling promotes differentiation of myeloid precursors, as well as the continued regulation of proliferation, survival and function of mononuclear phagocytes, dendritic cells and osteoclasts. While IL-34 and CSF-1 may induce similar cellular responses, they are differentially expressed and as such exert complimentary actions via CD115.The AFS98 antibody may be used for identification of myeloid lineage cells by flow cytometry, and is commonly used for in vivo or in vitro neutralization of CSF-1 Receptor (use format appropriate for functional assays).
Catalog No. 50-210-3416