Learn More
Biolegend PE anti-mouse CD127 (IL-7Rα)
Supplier:  Biolegend 158204
Encompass_Preferred
PE anti-mouse CD127 (IL-7Rα) [S18006K]; Isotype: Rat IgG2b, λ; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. 50-207-2395