Learn More
Cytek Biosciences Inc Antibodies PE Anti-Mouse CD2 (RM2-5) ..... Cat. No. 50-0021-U025 ..... Size 25 µg
Supplier:  Cytek Biosciences Inc 500021U025
Encompass_Preferred
Antibodies PE Anti-Mouse CD2 (RM2-5) ..... Cat. No. 50-0021-U025 ..... Size 25 µgThe RM2-5 antibody reacts with mouse CD2, an approximately 50 kDa glycoprotein, and a member of the Ig superfamily. CD2, also known as LFA-2, is a receptor for CD48 in the mouse and is expressed on the cell surface of all mouse lymphocytes. CD2 mediates adhesion between cells and is involved in T cell activation.RM2-5 is reported to block CD2-mediated cell:cell adhesion. Please choose the appropriate format for each application.
Catalog No. 50-210-3346