Learn More
Cytek Biosciences Inc Antibodies PE Anti-Mouse CD272 (BTLA) (6A6) ..... Cat. No. 50-5954-U025 ..... Size 25 µg
Supplier:  Cytek Biosciences Inc 505954U025
Encompass_Preferred
Antibodies PE Anti-Mouse CD272 (BTLA) (6A6) ..... Cat. No. 50-5954-U025 ..... Size 25 µgThe 6A6 antibody reacts with mouse CD272, a type 1 transmembrane glycoprotein and member of the Ig superfamily. CD272, also known as BTLA (B and T Lymphocyte Attenuator), is expressed on peripheral lymphocytes as well as developing B and T cells, macrophages, and dendritic cells. CD272 is a receptor for CD270 (HVEM) and is structurally similar to CD152 (CTLA-4) and CD279 (PD-1) and functions as a negative regulator of B and T cell responses.
Catalog No. 50-210-3448