Learn More
Biolegend PE anti-mouse CD274 (B7-H1, PD-L1)
Supplier:  Biolegend 155403
Encompass_Preferred
PE anti-mouse CD274 (B7-H1, PD-L1) [MIH7]; Isotype: Rat IgG2a, λ; Reactivity: Mouse; Apps: FC; Size: 25 ug
Catalog No. NC1565884