Learn More
Cytek Biosciences Inc PE aMu CD3 Anti Primary
Supplier: Cytek Biosciences Inc 500032U025
Product Name: PE Anti-Mouse CD3 (17A2)
Product Class: Antibody
Type: Primary
Product Name: PE Anti-Mouse CD3 (17A2)
Cytek Catalog Number: 50-0032-U025
Marker: CD3
Target Species: Mouse
Clonality: Monoclonal
Clone: 17A2
Host Species: Rat
Isotype: IgG2b, k
Conjugation: PE
Size: 25 μg
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.