Learn More
Biolegend PE anti-mouse CD304 (Neuropilin-1)
Supplier:  Biolegend 145203
Encompass_Preferred
PE anti-mouse CD304 (Neuropilin-1) [3E12]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 25 ug
Catalog No. 50-605-118