Learn More
Biolegend PE anti-mouse CD31 (PECAM-1)
Supplier:  Biolegend 160204
Encompass_Preferred
PE anti-mouse CD31 (PECAM-1) [W18222B]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. NC1862284